The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015906.3 | NP_056990.3 | 2 - 3 | 724 | 72 |
NM_033020.2 | NP_148980.2 | 2 - 3 | 724 | 72 | |
XM_005270936.3 | XP_005270993.1 | 2 - 3 | 645 | 72 | |
XM_005270937.3 | XP_005270994.1 | 2 - 3 | 645 | 72 | |
XM_011541568.2 | XP_011539870.1 | 2 - 3 | 645 | 72 | |
XM_017001452.1 | XP_016856941.1 | 1 - 2 | 114 | 72 | |
XM_017001453.1 | XP_016856942.1 | 1 - 2 | 114 | 72 | |
XM_017001454.1 | XP_016856943.1 | 2 - 3 | 645 | 72 | |
GenBank mRNA | AB029036.1 | - | 2 - 3 | 653 | 72 |
AF119043.1 | - | 2 - 3 | 724 | 72 | |
AF220136.1 | - | 2 - 3 | 724 | 72 | |
AF220137.1 | - | 2 - 3 | 724 | 72 | |
AJ132948.1 | - | 2 - 3 | 415 | 72 |