The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257374.2 | NP_001244303.1 | 4 - 5 | 562 | 83 |
NM_001282624.1 | NP_001269553.1 | 5 - 6 | 742 | 83 | |
NM_001282625.1 | NP_001269554.1 | 7 - 8 | 1484 | 83 | |
NM_001282626.1 | NP_001269555.1 | 4 - 5 | 1060 | 83 | |
NM_005572.3 | NP_005563.1 | 4 - 5 | 1060 | 83 | |
NM_170707.3 | NP_733821.1 | 4 - 5 | 1060 | 83 | |
NM_170708.3 | NP_733822.1 | 4 - 5 | 1060 | 83 | |
XM_011509533.1 | XP_011507835.1 | 4 - 5 | 557 | 83 | |
XM_011509534.1 | XP_011507836.1 | 3 - 4 | 455 | 83 | |
GenBank mRNA | AK026584.1 | - | 4 - 5 | 1014 | 83 |
AK056191.1 | - | 5 - 6 | 635 | 83 | |
AK057997.1 | - | 4 - 5 | 909 | 83 | |
AK097801.1 | - | 5 - 6 | 670 | 83 | |
AK098128.1 | - | 1 - 2 | 156 | 83 | |
AK122732.1 | - | 5 - 6 | 946 | 83 | |
AK130179.1 | - | 4 - 5 | 967 | 83 | |
AK295390.1 | - | 4 - 5 | 509 | 83 | |
AY357727.1 | - | 4 - 5 | 1008 | 83 | |
AY528714.1 | - | 4 - 5 | 929 | 83 | |
AY847595.1 | - | 4 - 5 | 811 | 83 | |
AY847596.1 | - | 4 - 5 | 811 | 83 | |
AY847597.1 | - | 4 - 5 | 811 | 83 | |
BC000511.2 | - | 4 - 5 | 991 | 83 | |
BC003162.1 | - | 4 - 5 | 976 | 83 | |
BC014507.1 | - | 4 - 5 | 1008 | 83 | |
BC033088.1 | - | 4 - 5 | 1004 | 83 | |
DC400775.1 | - | 4 - 5 | 519 | 83 | |
M13451.1 | - | 4 - 5 | 945 | 83 | |
M13452.1 | - | 3 - 4 | 364 | 83 | |
X03444.1 | - | 4 - 5 | 1021 | 83 | |
X03445.1 | - | 4 - 5 | 1021 | 83 |