The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018263.4 | NP_060733.4 | 5 - 6 | 624 | 83 |
XM_011532950.2 | XP_011531252.1 | 4 - 5 | 663 | 83 | |
XM_011532951.2 | XP_011531253.1 | 3 - 4 | 351 | 83 | |
XM_017004429.1 | XP_016859918.1 | 5 - 6 | 454 | 83 | |
GenBank mRNA | AB051472.2 | - | 5 - 6 | 624 | 83 |
AB084279.1 | - | 5 - 6 | 487 | 83 | |
BX647527.1 | - | 6 - 7 | 697 | 83 | |
BX648218.1 | - | 5 - 6 | 649 | 83 | |
CR933613.1 | - | 5 - 6 | 668 | 83 |