Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAB1
Entrez Gene ID
23166
Gene Name
stabilin 1
Gene Aliases
CLEVER-1, FEEL-1, FELE-1, FEX1, SCARH2, STAB-1
Chromosome Location
Chr.3: 52495338 - 52524496 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015136.2 NP_055951.2 49 - 50 5252 82
XM_005264973.3 XP_005265030.1 49 - 50 5254 82
XM_005264974.2 XP_005265031.1 49 - 50 5254 82
XM_017005998.1 XP_016861487.1 49 - 50 5314 82
XM_017005999.1 XP_016861488.1 49 - 50 5314 82
XM_017006000.1 XP_016861489.1 49 - 50 5314 82
XM_017006001.1 XP_016861490.1 48 - 49 5218 82
XM_017006002.1 XP_016861491.1 49 - 50 5314 82
XM_017006003.1 XP_016861492.1 49 - 50 5314 82
XM_017006004.1 XP_016861493.1 49 - 50 5314 82
GenBank mRNA AB052956.1 - 49 - 50 5239 82
AJ275213.1 - 49 - 50 5208 82
AK097061.1 - 8 - 9 1901 82
BC150250.1 - 49 - 50 5258 82
D87433.2 - 49 - 50 5237 82
DQ786774.1 - 49 - 50 5202 82

View Full Product Details