Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PTP4A1
- Entrez Gene ID
-
7803
- Gene Name
- protein tyrosine phosphatase type IVA, member 1
- Gene Aliases
-
HH72, PRL-1, PRL1, PTP(CAAX1), PTPCAAX1
- Chromosome Location
-
Chr.6: 63528021 - 63583588 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CR749458.1 |
- |
1 - 2 |
85 |
103 |
View Full Product Details