Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BSN
Entrez Gene ID
8927
Gene Name
bassoon presynaptic cytomatrix protein
Gene Aliases
ZNF231
Chromosome Location
Chr.3: 49554489 - 49671549 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003458.3 NP_003449.2 4 - 5 2099 81
GenBank mRNA AF052224.1 - 4 - 5 2100 81

View Full Product Details