The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015540.3 | NP_056355.2 | 16 - 17 | 2372 | 101 |
XM_005254297.1 | XP_005254354.1 | 16 - 17 | 2651 | 101 | |
XM_006720469.1 | XP_006720532.1 | 15 - 16 | 2186 | 101 | |
GenBank mRNA | AB037824.1 | - | 16 - 17 | 2340 | 101 |
AK022794.1 | - | 6 - 7 | 593 | 101 | |
AK290224.1 | - | 16 - 17 | 2353 | 101 | |
BC000246.1 | - | 16 - 17 | 2354 | 101 |