Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPAP1
Entrez Gene ID
26015
Gene Name
RNA polymerase II associated protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 41517176 - 41544277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015540.3 NP_056355.2 7 - 8 1079 70
XM_005254297.1 XP_005254354.1 7 - 8 1358 70
GenBank mRNA AB037824.1 - 7 - 8 1047 70
AK290224.1 - 7 - 8 1060 70
BC000246.1 - 7 - 8 1061 70

View Full Product Details