The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015540.3 | NP_056355.2 | 9 - 10 | 1298 | 72 |
XM_005254297.1 | XP_005254354.1 | 9 - 10 | 1577 | 72 | |
XM_006720469.1 | XP_006720532.1 | 8 - 9 | 1112 | 72 | |
GenBank mRNA | AB037824.1 | - | 9 - 10 | 1266 | 72 |
AK290224.1 | - | 9 - 10 | 1279 | 72 | |
BC000246.1 | - | 9 - 10 | 1280 | 72 |