Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC6A16
Entrez Gene ID
28968
Gene Name
solute carrier family 6 member 16
Gene Aliases
NT5, NTT5
Chromosome Location
Chr.19: 49289632 - 49332320 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014037.2 NP_054756.2 11 - 12 2175 70
XM_005258820.2 XP_005258877.1 12 - 13 2276 70
XM_006723168.3 XP_006723231.1 11 - 12 2191 70
XM_011526859.2 XP_011525161.2 10 - 11 2098 70
XM_011526860.2 XP_011525162.1 12 - 13 2488 70
XM_011526861.2 XP_011525163.1 10 - 11 1598 70
XM_017026712.1 XP_016882201.1 11 - 12 2191 70
GenBank mRNA AF265578.1 - 10 - 11 1941 70
AL136856.1 - 10 - 11 1987 70

View Full Product Details