Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MAST3
- Entrez Gene ID
-
23031
- Gene Name
- microtubule associated serine/threonine kinase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 18097764 - 18151692 on Build GRCh38
View Full Product Details