The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000552.4 | NP_000543.2 | 1 - 2 | 250 | 72 |
GenBank mRNA | AK292122.1 | - | 1 - 2 | 158 | 72 |
AK298097.1 | - | 1 - 2 | 245 | 72 | |
AK301216.1 | - | 1 - 2 | 118 | 72 | |
BC022258.1 | - | 1 - 2 | 245 | 72 | |
CD690980.1 | - | 1 - 2 | 277 | 72 | |
M17588.1 | - | 1 - 2 | 205 | 72 | |
X04146.1 | - | 1 - 2 | 224 | 72 | |
X04385.1 | - | 1 - 2 | 94 | 72 |