The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000552.4 | NP_000543.2 | 20 - 21 | 2938 | 103 |
GenBank mRNA | AF086470.1 | - | 3 - 4 | 357 | 103 |
M10321.1 | - | 4 - 5 | 467 | 103 | |
X04146.1 | - | 20 - 21 | 2912 | 103 | |
X04385.1 | - | 20 - 21 | 2782 | 103 |