Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWF
Entrez Gene ID
7450
Gene Name
von Willebrand factor
Gene Aliases
F8VWF, VWD
Chromosome Location
Chr.12: 5948874 - 6124675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000552.4 NP_000543.2 29 - 30 5427 85
GenBank mRNA AK297600.1 - 1 - 2 56 85
M10321.1 - 13 - 14 2956 85
X04385.1 - 29 - 30 5271 85

View Full Product Details