Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWF
Entrez Gene ID
7450
Gene Name
von Willebrand factor
Gene Aliases
F8VWF, VWD
Chromosome Location
Chr.12: 5948874 - 6124675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000552.4 NP_000543.2 38 - 39 7051 115
GenBank mRNA AK297600.1 - 10 - 11 1680 115
M10321.1 - 22 - 23 4580 115
X04385.1 - 38 - 39 6895 115

View Full Product Details