The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000552.4 | NP_000543.2 | 39 - 40 | 7156 | 76 |
GenBank mRNA | AK297600.1 | - | 11 - 12 | 1785 | 76 |
BQ898017.1 | - | 1 - 2 | 83 | 76 | |
M10321.1 | - | 23 - 24 | 4685 | 76 | |
X04385.1 | - | 39 - 40 | 7000 | 76 |