Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWF
Entrez Gene ID
7450
Gene Name
von Willebrand factor
Gene Aliases
F8VWF, VWD
Chromosome Location
Chr.12: 5948874 - 6124675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000552.4 NP_000543.2 47 - 48 8144 91
GenBank mRNA AK297600.1 - 19 - 20 2773 91
M10321.1 - 31 - 32 5673 91
X04385.1 - 47 - 48 7988 91

View Full Product Details