The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005159.4 | NP_005150.1 | 3 - 4 | 696 | 70 |
GenBank mRNA | AK129892.1 | - | 3 - 4 | 519 | 70 |
AK129902.1 | - | 3 - 4 | 519 | 70 | |
AK290616.1 | - | 3 - 4 | 519 | 70 | |
BC009978.2 | - | 3 - 4 | 658 | 70 | |
CR541795.1 | - | 2 - 3 | 453 | 70 |