The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110.3 | NP_001101.1 | 6 - 7 | 1176 | 186 |
XM_017021861.1 | XP_016877350.1 | 7 - 8 | 1231 | 186 | |
GenBank mRNA | AF009615.1 | - | 6 - 7 | 1201 | 186 |
BC126253.1 | - | 6 - 7 | 1032 | 186 | |
BC143935.1 | - | 6 - 7 | 1032 | 186 | |
Z48579.1 | - | 4 - 5 | 561 | 186 |