The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110.3 | NP_001101.1 | 3 - 4 | 766 | 148 |
NM_001320570.1 | NP_001307499.1 | 3 - 4 | 766 | 148 | |
XM_017021861.1 | XP_016877350.1 | 4 - 5 | 821 | 148 | |
GenBank mRNA | AF009615.1 | - | 3 - 4 | 791 | 148 |
BC126253.1 | - | 3 - 4 | 622 | 148 | |
BC143935.1 | - | 3 - 4 | 622 | 148 |