Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRNP200
Entrez Gene ID
23020
Gene Name
small nuclear ribonucleoprotein U5 subunit 200
Gene Aliases
ASCC3L1, BRR2, HELIC2, RP33, U5-200KD
Chromosome Location
Chr.2: 96274336 - 96321893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014014.4 NP_054733.2 17 - 18 2439 69
XM_017003603.1 XP_016859092.1 19 - 20 3269 69
GenBank mRNA AB018331.2 - 15 - 16 1977 69
AK024391.1 - 11 - 12 1520 69
AL831994.1 - 8 - 9 1155 69
AY572488.1 - 17 - 18 2385 69
BC065924.1 - 10 - 11 1362 69
BC112891.1 - 11 - 12 1560 69

View Full Product Details