The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000425.4 | NP_000416.1 | 12 - 13 | 1759 | 104 |
NM_001143963.2 | NP_001137435.1 | 11 - 12 | 1643 | 104 | |
NM_001278116.1 | NP_001265045.1 | 13 - 14 | 1768 | 104 | |
NM_024003.3 | NP_076493.1 | 12 - 13 | 1658 | 104 | |
GenBank mRNA | AB102653.1 | - | 12 - 13 | 1544 | 104 |
AK289754.1 | - | 12 - 13 | 1575 | 104 | |
AY927629.1 | - | 7 - 8 | 975 | 104 | |
BC126229.1 | - | 12 - 13 | 1660 | 104 | |
BC136447.1 | - | 12 - 13 | 1660 | 104 | |
M74387.1 | - | 12 - 13 | 1561 | 104 | |
M77640.1 | - | 12 - 13 | 1570 | 104 | |
X58776.1 | - | 4 - 5 | 494 | 104 | |
X59847.1 | - | 12 - 13 | 1550 | 104 |