The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000425.4 | NP_000416.1 | 5 - 6 | 730 | 62 |
NM_001143963.2 | NP_001137435.1 | 4 - 5 | 614 | 62 | |
NM_001278116.1 | NP_001265045.1 | 6 - 7 | 739 | 62 | |
NM_024003.3 | NP_076493.1 | 5 - 6 | 629 | 62 | |
GenBank mRNA | AB102653.1 | - | 5 - 6 | 515 | 62 |
AK289754.1 | - | 5 - 6 | 546 | 62 | |
BC126229.1 | - | 5 - 6 | 631 | 62 | |
BC136447.1 | - | 5 - 6 | 631 | 62 | |
EF506611.1 | - | 4 - 5 | 506 | 62 | |
M74387.1 | - | 5 - 6 | 532 | 62 | |
M77640.1 | - | 5 - 6 | 541 | 62 | |
X59847.1 | - | 5 - 6 | 521 | 62 |