The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277161.1 | NP_001264090.1 | 10 - 11 | 2799 | 66 |
NM_004745.4 | NP_004736.2 | 11 - 12 | 2841 | 66 | |
GenBank mRNA | AB000275.1 | - | 8 - 9 | 1396 | 66 |
AF009204.1 | - | 9 - 10 | 2612 | 66 | |
AF119817.1 | - | 11 - 12 | 2826 | 66 | |
AF119818.1 | - | 11 - 12 | 2841 | 66 | |
BC141862.1 | - | 10 - 11 | 2757 | 66 | |
KJ534823.1 | - | 10 - 11 | 2707 | 66 |