The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277161.1 | NP_001264090.1 | 2 - 3 | 1119 | 101 |
NM_004745.4 | NP_004736.2 | 2 - 3 | 1119 | 101 | |
GenBank mRNA | AF009204.1 | - | 1 - 2 | 932 | 101 |
AF119817.1 | - | 2 - 3 | 1104 | 101 | |
AF119818.1 | - | 2 - 3 | 1119 | 101 | |
BC141862.1 | - | 1 - 2 | 1035 | 101 | |
KJ534823.1 | - | 1 - 2 | 985 | 101 |