Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOL9
Entrez Gene ID
79707
Gene Name
nucleolar protein 9
Gene Aliases
Grc3, NET6
Chromosome Location
Chr.1: 6521347 - 6554598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024654.4 NP_078930.3 2 - 3 711 88
XM_005263493.3 XP_005263550.1 1 - 2 99 88
XM_017002337.1 XP_016857826.1 2 - 3 649 88
XM_017002338.1 XP_016857827.1 2 - 3 454 88
XM_017002339.1 XP_016857828.1 2 - 3 268 88
GenBank mRNA AK091284.1 - 2 - 3 648 88
BC094836.1 - 1 - 2 86 88
BC105095.1 - 2 - 3 711 88
BC110849.1 - 2 - 3 620 88
BC112278.1 - 2 - 3 711 88
BC143646.1 - 2 - 3 711 88
BI828678.1 - 2 - 3 613 88

View Full Product Details