The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040142.1 | NP_001035232.1 | 10 - 11 | 1667 | 91 |
NM_001040143.1 | NP_001035233.1 | 9 - 10 | 1428 | 91 | |
NM_021007.2 | NP_066287.2 | 10 - 11 | 1733 | 91 | |
XM_005246753.3 | XP_005246810.1 | 10 - 11 | 1683 | 91 | |
XM_005246755.4 | XP_005246812.1 | 5 - 6 | 768 | 91 | |
XM_017004655.1 | XP_016860144.1 | 10 - 11 | 1582 | 91 | |
XM_017004656.1 | XP_016860145.1 | 10 - 11 | 1638 | 91 | |
XM_017004657.1 | XP_016860146.1 | 10 - 11 | 1535 | 91 | |
XM_017004658.1 | XP_016860147.1 | 10 - 11 | 1847 | 91 | |
GenBank mRNA | AB208888.1 | - | 10 - 11 | 1520 | 91 |
AK289656.1 | - | 10 - 11 | 1667 | 91 | |
BC029489.1 | - | 10 - 11 | 1624 | 91 | |
BC047398.1 | - | 10 - 11 | 1624 | 91 | |
BC048417.1 | - | 10 - 11 | 1624 | 91 | |
M94055.1 | - | 10 - 11 | 1482 | 91 |