Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN2A
Entrez Gene ID
6326
Gene Name
sodium voltage-gated channel alpha subunit 2
Gene Aliases
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2
Chromosome Location
Chr.2: 165208056 - 165392310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040142.1 NP_001035232.1 1 - 2 236 137
XM_005246753.3 XP_005246810.1 1 - 2 252 137
XM_017004658.1 XP_016860147.1 1 - 2 229 137
GenBank mRNA AK289656.1 - 1 - 2 236 137
BC029489.1 - 1 - 2 193 137
BC047398.1 - 1 - 2 193 137
BC048417.1 - 1 - 2 193 137
BP199510.1 - 1 - 2 236 137

View Full Product Details