The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040142.1 | NP_001035232.1 | 4 - 5 | 762 | 107 |
NM_001040143.1 | NP_001035233.1 | 3 - 4 | 523 | 107 | |
NM_021007.2 | NP_066287.2 | 4 - 5 | 828 | 107 | |
XM_005246753.3 | XP_005246810.1 | 4 - 5 | 778 | 107 | |
XM_017004655.1 | XP_016860144.1 | 4 - 5 | 677 | 107 | |
XM_017004656.1 | XP_016860145.1 | 4 - 5 | 733 | 107 | |
XM_017004657.1 | XP_016860146.1 | 4 - 5 | 630 | 107 | |
XM_017004658.1 | XP_016860147.1 | 4 - 5 | 755 | 107 | |
GenBank mRNA | AB208888.1 | - | 4 - 5 | 615 | 107 |
AK289656.1 | - | 4 - 5 | 762 | 107 | |
BC029489.1 | - | 4 - 5 | 719 | 107 | |
BC047398.1 | - | 4 - 5 | 719 | 107 | |
BC048417.1 | - | 4 - 5 | 719 | 107 | |
M94055.1 | - | 4 - 5 | 577 | 107 |