Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN2A
Entrez Gene ID
6326
Gene Name
sodium voltage-gated channel alpha subunit 2
Gene Aliases
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2
Chromosome Location
Chr.2: 165208056 - 165392310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040142.1 NP_001035232.1 7 - 8 1251 118
NM_001040143.1 NP_001035233.1 6 - 7 1012 118
NM_021007.2 NP_066287.2 7 - 8 1317 118
XM_005246753.3 XP_005246810.1 7 - 8 1267 118
XM_005246755.4 XP_005246812.1 2 - 3 352 118
XM_017004655.1 XP_016860144.1 7 - 8 1166 118
XM_017004656.1 XP_016860145.1 7 - 8 1222 118
XM_017004657.1 XP_016860146.1 7 - 8 1119 118
XM_017004658.1 XP_016860147.1 7 - 8 1431 118
GenBank mRNA AB208888.1 - 7 - 8 1104 118
AK289656.1 - 7 - 8 1251 118
BC029489.1 - 7 - 8 1208 118
BC047398.1 - 7 - 8 1208 118
BC048417.1 - 7 - 8 1208 118
M94055.1 - 7 - 8 1066 118

View Full Product Details