Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNL1
Entrez Gene ID
2794
Gene Name
G protein nucleolar 1 (putative)
Gene Aliases
HSR1
Chromosome Location
Chr.6: 30541377 - 30557594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005275.3 NP_005266.2 12 - 12 6728 71
XM_005249015.3 XP_005249072.1 13 - 13 6191 71
GenBank mRNA AK095071.1 - 1 - 1 1923 71

View Full Product Details