Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACE
Entrez Gene ID
1636
Gene Name
angiotensin I converting enzyme
Gene Aliases
ACE1, CD143, DCP, DCP1, ICH, MVCD3
Chromosome Location
Chr.17: 63477061 - 63498380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000789.3 NP_000780.1 12 - 13 1955 83
GenBank mRNA AB208971.1 - 12 - 13 1945 83
J04144.1 - 12 - 13 1943 83
JQ927553.1 - 12 - 13 1955 83

View Full Product Details