The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000065.3 | NP_000056.2 | 2 - 3 | 381 | 108 |
NM_001115131.2 | NP_001108603.2 | 2 - 3 | 408 | 108 | |
XM_005248357.2 | XP_005248414.1 | 2 - 3 | 456 | 108 | |
XM_006714496.3 | XP_006714559.1 | 2 - 3 | 456 | 108 | |
XM_011514114.2 | XP_011512416.1 | 2 - 3 | 456 | 108 | |
XM_011514115.2 | XP_011512417.1 | 2 - 3 | 377 | 108 | |
XM_011514116.2 | XP_011512418.1 | 3 - 4 | 493 | 108 | |
XM_011514117.2 | XP_011512419.1 | 3 - 4 | 535 | 108 | |
XM_011514118.2 | XP_011512420.1 | 2 - 3 | 273 | 108 | |
XM_011514119.2 | XP_011512421.1 | 2 - 3 | 456 | 108 | |
XM_017009818.1 | XP_016865307.1 | 3 - 4 | 516 | 108 | |
GenBank mRNA | AB126592.1 | - | 1 - 2 | 143 | 108 |
AK292509.1 | - | 2 - 3 | 279 | 108 | |
AK313673.1 | - | 2 - 3 | 300 | 108 | |
BC035723.1 | - | 2 - 3 | 408 | 108 | |
DB066614.1 | - | 2 - 3 | 381 | 108 | |
DB184605.1 | - | 2 - 3 | 353 | 108 | |
J04506.1 | - | 2 - 3 | 209 | 108 | |
J05024.1 | - | 2 - 3 | 200 | 108 | |
J05064.1 | - | 2 - 3 | 298 | 108 |