Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC30A8
Entrez Gene ID
169026
Gene Name
solute carrier family 30 member 8
Gene Aliases
ZNT8, ZnT-8
Chromosome Location
Chr.8: 116950273 - 117176714 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172811.1 NP_001166282.1 2 - 3 212 88
GenBank mRNA EF560713.1 - 2 - 3 212 88

View Full Product Details