The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152701.4 | NP_689914.3 | 59 - 60 | 14861 | 72 |
XM_011515130.2 | XP_011513432.1 | 59 - 60 | 14896 | 72 | |
XM_011515131.2 | XP_011513433.1 | 59 - 60 | 14803 | 72 | |
XM_011515132.2 | XP_011513434.1 | 58 - 59 | 14803 | 72 | |
XM_011515133.2 | XP_011513435.1 | 58 - 59 | 14758 | 72 | |
XM_011515136.2 | XP_011513438.2 | 54 - 55 | 14261 | 72 | |
GenBank mRNA | AF501281.1 | - | 41 - 42 | 6044 | 72 |
AK091270.1 | - | 7 - 8 | 864 | 72 | |
AK128516.1 | - | 16 - 17 | 2156 | 72 | |
AK131360.1 | - | 27 - 28 | 4067 | 72 | |
AY204751.1 | - | 59 - 60 | 14862 | 72 | |
JF913488.1 | - | 42 - 43 | 6759 | 72 | |
JF913491.1 | - | 42 - 43 | 6666 | 72 |