Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
Chromosome Location
Chr.7: 48171424 - 48647497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152701.4 NP_689914.3 10 - 11 1290 82
XM_011515130.2 XP_011513432.1 10 - 11 1322 82
XM_011515131.2 XP_011513433.1 10 - 11 1322 82
XM_011515132.2 XP_011513434.1 10 - 11 1322 82
XM_011515133.2 XP_011513435.1 10 - 11 1322 82
XM_011515134.2 XP_011513436.1 10 - 11 1322 82
XM_011515136.2 XP_011513438.2 5 - 6 687 82
XM_011515137.2 XP_011513439.1 10 - 11 1322 82
XM_011515138.2 XP_011513440.1 10 - 11 1322 82
XM_011515139.2 XP_011513441.1 10 - 11 1322 82
XM_011515141.2 XP_011513443.1 10 - 11 1323 82
XM_011515142.2 XP_011513444.1 10 - 11 1323 82
XM_011515143.2 XP_011513445.1 10 - 11 1322 82
XM_011515144.2 XP_011513446.1 10 - 11 1322 82
XM_011515145.2 XP_011513447.1 10 - 11 1323 82
XM_011515146.2 XP_011513448.1 10 - 11 1323 82
XM_011515147.2 XP_011513449.1 10 - 11 1324 82
XM_011515148.2 XP_011513450.1 10 - 11 1325 82
XM_011515149.2 XP_011513451.1 10 - 11 1325 82
XM_017011767.1 XP_016867256.1 10 - 11 1322 82
XM_017011768.1 XP_016867257.1 10 - 11 1322 82
XM_017011769.1 XP_016867258.1 10 - 11 1326 82
GenBank mRNA AY204751.1 - 10 - 11 1291 82

View Full Product Details