The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318517.1 | NP_001305446.1 | 2 - 3 | 302 | 122 |
NM_001318518.1 | NP_001305447.1 | 2 - 3 | 302 | 122 | |
XM_006719992.1 | XP_006720055.1 | 2 - 3 | 292 | 122 | |
GenBank mRNA | AF164598.1 | - | 2 - 3 | 108 | 122 |
AK315302.1 | - | 2 - 3 | 136 | 122 | |
BC010875.2 | - | 2 - 3 | 209 | 122 | |
U18291.1 | - | 2 - 3 | 128 | 122 |