The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005687.4 | NP_005678.3 | 2 - 3 | 398 | 83 |
GenBank mRNA | AF042346.1 | - | 2 - 3 | 116 | 83 |
AF161521.1 | - | 2 - 3 | 136 | 83 | |
AK291531.1 | - | 2 - 3 | 133 | 83 | |
BC006502.2 | - | 2 - 3 | 105 | 83 | |
BC017783.1 | - | 2 - 3 | 128 | 83 | |
BG721840.1 | - | 2 - 3 | 400 | 83 | |
D84430.1 | - | 2 - 3 | 129 | 83 |