The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005687.4 | NP_005678.3 | 5 - 6 | 736 | 87 |
NR_130154.1 | 6 - 7 | 932 | 87 | ||
XM_006712169.2 | XP_006712232.1 | 6 - 7 | 638 | 87 | |
XM_011510466.2 | XP_011508768.1 | 6 - 7 | 510 | 87 | |
XM_017003110.1 | XP_016858599.1 | 6 - 7 | 2911 | 87 | |
GenBank mRNA | AF042346.1 | - | 5 - 6 | 454 | 87 |
AF161521.1 | - | 5 - 6 | 474 | 87 | |
AK055289.1 | - | 3 - 4 | 1700 | 87 | |
AK291531.1 | - | 5 - 6 | 471 | 87 | |
AK294158.1 | - | 6 - 7 | 492 | 87 | |
AK316497.1 | - | 5 - 6 | 361 | 87 | |
BC006502.2 | - | 5 - 6 | 443 | 87 | |
BC017783.1 | - | 5 - 6 | 466 | 87 | |
D84430.1 | - | 5 - 6 | 467 | 87 |