The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005687.4 | NP_005678.3 | 7 - 8 | 1002 | 100 |
NR_130154.1 | 8 - 9 | 1198 | 100 | ||
XM_006712169.2 | XP_006712232.1 | 8 - 9 | 904 | 100 | |
XM_011510466.2 | XP_011508768.1 | 8 - 9 | 776 | 100 | |
XM_017003110.1 | XP_016858599.1 | 8 - 9 | 3177 | 100 | |
GenBank mRNA | AF042346.1 | - | 7 - 8 | 720 | 100 |
AF161521.1 | - | 7 - 8 | 740 | 100 | |
AK055289.1 | - | 5 - 6 | 1966 | 100 | |
AK294158.1 | - | 8 - 9 | 758 | 100 | |
AK316497.1 | - | 7 - 8 | 627 | 100 | |
BC006502.2 | - | 7 - 8 | 709 | 100 | |
BC017783.1 | - | 7 - 8 | 732 | 100 | |
D84430.1 | - | 7 - 8 | 733 | 100 |