Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF25
Entrez Gene ID
64320
Gene Name
ring finger protein 25
Gene Aliases
AO7
Chromosome Location
Chr.2: 218663864 - 218672058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022453.2 NP_071898.2 5 - 6 442 73
XM_017004695.1 XP_016860184.1 5 - 6 442 73
GenBank mRNA AK023968.1 - 5 - 6 360 73
AK222525.1 - 5 - 6 380 73
AK289501.1 - 5 - 6 386 73
BC015612.2 - 5 - 6 359 73
BI831198.1 - 5 - 6 442 73

View Full Product Details