Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191028.2 NP_001177957.2 18 - 19 2634 103
NM_012470.3 NP_036602.1 19 - 20 2826 103
NR_034053.2 20 - 21 2987 103
XM_011515989.2 XP_011514291.1 19 - 20 2428 103
GenBank mRNA AF145029.1 - 19 - 20 2638 103
AJ133769.1 - 19 - 20 2423 103
AK001263.1 - 12 - 13 1380 103
AK022910.1 - 10 - 11 1160 103
AK225999.1 - 18 - 19 2605 103
AK312580.1 - 19 - 20 2553 103
BC009221.2 - 7 - 8 670 103
BC068530.1 - 19 - 20 2554 103

View Full Product Details