Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191028.2 NP_001177957.2 3 - 4 804 88
NM_012470.3 NP_036602.1 3 - 4 804 88
NR_034053.2 4 - 5 965 88
XM_011515989.2 XP_011514291.1 3 - 4 406 88
GenBank mRNA AF145029.1 - 3 - 4 514 88
AJ133769.1 - 3 - 4 401 88
AK225999.1 - 3 - 4 775 88
AK312580.1 - 3 - 4 531 88
BC068530.1 - 3 - 4 532 88

View Full Product Details