Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191028.2 NP_001177957.2 4 - 5 959 106
NM_012470.3 NP_036602.1 4 - 5 959 106
NR_034053.2 5 - 6 1120 106
XM_011515989.2 XP_011514291.1 4 - 5 561 106
GenBank mRNA AF145029.1 - 4 - 5 669 106
AJ133769.1 - 4 - 5 556 106
AK225999.1 - 4 - 5 930 106
AK312580.1 - 4 - 5 686 106
BC068530.1 - 4 - 5 687 106

View Full Product Details