The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191028.2 | NP_001177957.2 | 10 - 11 | 1753 | 92 |
NM_012470.3 | NP_036602.1 | 10 - 11 | 1753 | 92 | |
NR_034053.2 | 11 - 12 | 1914 | 92 | ||
XM_011515989.2 | XP_011514291.1 | 10 - 11 | 1355 | 92 | |
GenBank mRNA | AJ133769.1 | - | 10 - 11 | 1350 | 92 |
AK001263.1 | - | 3 - 4 | 307 | 92 | |
AK022910.1 | - | 1 - 2 | 87 | 92 | |
AK225999.1 | - | 10 - 11 | 1724 | 92 | |
BC068530.1 | - | 10 - 11 | 1481 | 92 |