Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
Chromosome Location
Chr.14: 23412738 - 23435686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000257.3 NP_000248.2 10 - 11 1028 123
XM_017021340.1 XP_016876829.1 9 - 10 963 123
GenBank mRNA AK307590.1 - 10 - 11 1002 123
BC112171.1 - 9 - 10 952 123
BC112173.1 - 9 - 10 952 123
EF560725.1 - 8 - 9 895 123
EU747717.1 - 10 - 11 1003 123
M58018.1 - 10 - 11 983 123

View Full Product Details