Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUPT16H
Entrez Gene ID
11198
Gene Name
SPT16 homolog, facilitates chromatin remodeling subunit
Gene Aliases
CDC68, FACTP140, SPT16, SPT16/CDC68
Chromosome Location
Chr.14: 21351472 - 21384266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007192.3 NP_009123.1 3 - 4 659 109
XM_011536381.2 XP_011534683.1 4 - 5 661 109
GenBank mRNA AK024072.1 - 3 - 4 661 109
BC000565.2 - 3 - 4 396 109
BC014046.2 - 3 - 4 396 109
BC064561.1 - 3 - 4 384 109
BC073849.1 - 3 - 4 393 109
BC111402.1 - 3 - 4 408 109

View Full Product Details