The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290033.1 | NP_001276962.1 | 5 - 6 | 417 | 113 |
NM_012133.5 | NP_036265.3 | 5 - 6 | 417 | 113 | |
GenBank mRNA | AB047847.1 | - | 5 - 6 | 327 | 113 |
AF157833.1 | - | 5 - 6 | 387 | 113 | |
AF207598.1 | - | 5 - 6 | 383 | 113 | |
AK022934.1 | - | 5 - 6 | 314 | 113 | |
AK291409.1 | - | 5 - 6 | 407 | 113 | |
BC017443.1 | - | 5 - 6 | 310 | 113 | |
BC110796.1 | - | 5 - 6 | 364 | 113 | |
BG470623.1 | - | 5 - 6 | 308 | 113 | |
BG704011.1 | - | 5 - 6 | 383 | 113 | |
BX353590.2 | - | 5 - 6 | 417 | 113 |