The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135175.1 | NP_001128647.1 | 12 - 13 | 1819 | 98 |
NM_001135176.1 | NP_001128648.1 | 12 - 13 | 1738 | 98 | |
NM_001135177.1 | NP_001128649.1 | 11 - 12 | 1752 | 98 | |
NM_152905.3 | NP_690869.1 | 13 - 14 | 1991 | 98 | |
XM_005268644.2 | XP_005268701.1 | 13 - 14 | 1725 | 98 | |
XM_006719237.3 | XP_006719300.1 | 11 - 12 | 1481 | 98 | |
XM_011537903.2 | XP_011536205.1 | 10 - 11 | 1489 | 98 | |
XM_011537904.2 | XP_011536206.1 | 13 - 14 | 1814 | 98 | |
XM_011537905.2 | XP_011536207.1 | 12 - 13 | 1620 | 98 | |
XM_011537906.2 | XP_011536208.1 | 12 - 13 | 1615 | 98 | |
XM_017018801.1 | XP_016874290.1 | 12 - 13 | 1863 | 98 | |
XM_017018802.1 | XP_016874291.1 | 12 - 13 | 1858 | 98 | |
GenBank mRNA | AK093221.1 | - | 13 - 14 | 1929 | 98 |
AK290058.1 | - | 13 - 14 | 1991 | 98 | |
AK303656.1 | - | 11 - 12 | 1718 | 98 | |
AK315821.1 | - | 13 - 14 | 1957 | 98 | |
AL832466.1 | - | 12 - 13 | 1705 | 98 | |
AL832468.1 | - | 10 - 11 | 1664 | 98 | |
BC027605.2 | - | 12 - 13 | 1819 | 98 |