The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003401.4 | NP_003392.1 | 7 - 8 | 1108 | 84 |
NM_022550.3 | NP_072044.1 | 7 - 8 | 1155 | 84 | |
GenBank mRNA | U40622.1 | - | 7 - 8 | 941 | 84 |